Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine 98,0

Supplier: TCI America

Synonyms: Boc-Asn(Trt)-OH, Nɑ-Boc-Nγ-trityl-L-asparagine

B3789-25G B3789-5G
TCB3789-25GEA 507.18 CAD
TCB3789-25G TCB3789-5G
Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine 98,0
Nalpha-tert-Butoxycarbonyl-N-gamma-trityl-L-asparagine

CAS Number: 132388-68-2
MDL Number: MFCD00153299
Molecular Formula: C28H30N2O5
Molecular Weight: 474.56
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202
Specific rotation [a]20/D: -21 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-Ngamma-trityl-L-asparagine, Boc-Asn(Trt)-OH

Formula: C₂₈H₃₀N₂O₅
MW: 474.56 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00153299
CAS Number: 132388-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f53b680ccf6e8bd68e99b2aef95f9929fc341fd&vid=5f53b680ccf6e8bd68e99b2aef95f9929fc341fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR