tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(Hydroxymethyl)piperidine, N-BOC protected

B3787-25G B3787-5G
TCB3787-25GEA 218.04 CAD
TCB3787-25G TCB3787-5G
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate

CAS Number: 116574-71-1
MDL Number: MFCD03094733
Molecular Formula: C11H21NO3
Molecular Weight: 215.29
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: 1-Boc-3-(hydroxymethyl)piperidine, 1-Boc-3-piperidinemethanol, 1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)piperidine

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Melting Pt: 78 °C
MDL Number: MFCD03094733
CAS Number: 116574-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=377c3ff06009a4046d035b12c6713a5bd6f68d81&vid=377c3ff06009a4046d035b12c6713a5bd6f68d81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR