4-Bromo-2-(trifluoromethoxy)aniline ≥98.0% (by GC)
Supplier: TCI America
B3786-25G
B3786-5G
TCB3786-25GEA
93.22
CAD
TCB3786-25G
TCB3786-5G
4-Bromo-2-(trifluoromethoxy)aniline ≥98.0% (by GC)
4-Bromo-2-(trifluoromethoxy)aniline
CAS Number: 175278-09-8
MDL Number: MFCD00179338
Molecular Formula: C7H5BrF3NO
Molecular Weight: 256.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 110
Specific Gravity (20/20): 1.70
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Amino-4-bromo-2-(trifluoromethoxy)benzene
Formula:
C₇H₅BrF₃NO Boiling Pt: 115 °C Density: 1.70 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00179338 CAS Number: 175278-09-8 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...