5-Bromo-1H-indazole ≥97.0% (by GC)

Supplier: TCI America

B3785-1G B3785-5G
TCB3785-1GEA 88.48 CAD
TCB3785-1G TCB3785-5G
5-Bromo-1H-indazole ≥97.0% (by GC)
5-Bromo-1H-indazole

CAS Number: 53857-57-1
MDL Number: MFCD00839493
Molecular Formula: C7H5BrN2
Molecular Weight: 197.04
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 132

Formula: C₇H₅BrN₂
Melting Pt: 132 °C
MDL Number: MFCD00839493
CAS Number: 53857-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5be616c9303a4e741fd5fef58bb604c58c536ab&vid=e5be616c9303a4e741fd5fef58bb604c58c536ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR