4-Bromo-α,α,α,3-tetrafluorotoluene ≥98.0% (by GC)

Supplier: TCI America

B3783-25G B3783-5G
TCB3783-25GEA 276.5 CAD
TCB3783-25G TCB3783-5G
4-Bromo-α,α,α,3-tetrafluorotoluene ≥98.0% (by GC)
4-Bromo-α,α,α,3-tetrafluorotoluene

CAS Number: 40161-54-4
MDL Number: MFCD00042497
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Specific Gravity (20/20): 1.72
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-2-fluoro-4-(trifluoromethyl)benzene, 4-Bromo-alpha,alpha,alpha,3-tetrafluorotoluene

Formula: C₇H₃BrF₄
Boiling Pt: 145 °C
Density: 1.72 (20 °C)
MDL Number: MFCD00042497
CAS Number: 40161-54-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d858b16e0bfb5797feba8836d9e9ad988a81b46&vid=3d858b16e0bfb5797feba8836d9e9ad988a81b46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR