3-Bromo-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3782-1G B3782-5G
TCB3782-1GEA 34.76 CAD
TCB3782-1G TCB3782-5G
3-Bromo-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-5-fluorobenzoic acid

CAS Number: 176548-70-2
MDL Number: MFCD01569540
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 144

Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 144 °C
MDL Number: MFCD01569540
CAS Number: 176548-70-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ee8b68dd4393ba3acd8a5008086295022ce0a15&vid=5ee8b68dd4393ba3acd8a5008086295022ce0a15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR