5-Bromo-2,2-difluoro-1,3-benzodioxole ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole

B3781-25G B3781-5G
TCB3781-25GEA 325.48 CAD
TCB3781-25G TCB3781-5G
5-Bromo-2,2-difluoro-1,3-benzodioxole ≥97.0% (by GC)
5-Bromo-2,2-difluoro-1,3-benzodioxole

CAS Number: 33070-32-5
MDL Number: MFCD00236212
Molecular Formula: C7H3BrF2O2
Molecular Weight: 237.00
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Specific Gravity (20/20): 1.74


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Bromo-1,2-(difluoromethylenedioxy)benzene

Formula: C₇H₃BrF₂O₂
MW: 237 g/mol
Boiling Pt: 79 °C
Density: 1.74 (20 °C)
MDL Number: MFCD00236212
CAS Number: 33070-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96c4a2b41fc07632f5d174677902251b96d70e34&vid=96c4a2b41fc07632f5d174677902251b96d70e34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR