5-Bromo-2-fluoro-3-methylpyridine ≥98.0% (by GC)

Supplier: TCI America

B3775-1G B3775-5G
TCB3775-1GEA 63.2 CAD
TCB3775-1G TCB3775-5G
5-Bromo-2-fluoro-3-methylpyridine ≥98.0% (by GC)
5-Bromo-2-fluoro-3-methylpyridine

CAS Number: 29312-98-9
MDL Number: MFCD03095091
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Bromo-2-fluoro-3-picoline

Formula: C₆H₅BrFN
Melting Pt: 65 °C
MDL Number: MFCD03095091
CAS Number: 29312-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22b7fa2b2016008247532ecc1add0b0c637705c2&vid=22b7fa2b2016008247532ecc1add0b0c637705c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR