Butyl-4-hydroxybenzoate ≥99.0% (by GC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

B3771-25G B3771-5G
TCB3771-25GEA 72.14 CAD
TCB3771-25G TCB3771-5G
Butyl-4-hydroxybenzoate ≥99.0% (by GC, titration analysis) for biochemical research
Butyl 4-hydroxybenzoate

[for Biochemical Research]
CAS Number: 94-26-8
MDL Number: MFCD00016478
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 156
Melting point (°C): 70


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: Butylparaben, 4-Hydroxybenzoic Acid Butyl Ester

Formula: C₁₁H₁₄O₃
Boiling Pt: 156 °C
Melting Pt: 70 °C
MDL Number: MFCD00016478
CAS Number: 94-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29bbc8d48629d1c2a3a77c48cb28fc923d3f199f&vid=1f2588ec19e5b3fd99566c94b3c2486059bb1183&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR