(±)-1,3-Butanediol ≥99.0% (by GC) for biochemical research

Supplier: TCI America

B3770-25G B3770-5G
TCB3770-25GEA 82.16 CAD
TCB3770-25G TCB3770-5G
(±)-1,3-Butanediol ≥99.0% (by GC) for biochemical research
(±)-1,3-Butanediol

[for Biochemical Research]
CAS Number: 107-88-0
MDL Number: MFCD00004554
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 122
Specific Gravity (20/20): 1.01


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 1,3-Butylene Glycol, 1,3-Dihydroxybutane

Formula: C₄H₁₀O₂
Boiling Pt: 207 °C
Density: 1.01 (20 °C)
Flash Pt: 122 °C
MDL Number: MFCD00004554
CAS Number: 107-88-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78dd59975a1fa3752dce458220271cf0afc90512&vid=ab8d08d4e43268c4e51532e3cd08908245fdcfbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR