2-Bromo-5-iodopyridine ≥97.0%

Supplier: TCI America

B3765-25G B3765-5G
TCB3765-25GEA 139.04 CAD
TCB3765-25G TCB3765-5G
2-Bromo-5-iodopyridine ≥97.0%
2-Bromo-5-iodopyridine

CAS Number: 73290-22-9
MDL Number: MFCD03095201
Molecular Formula: C5H3BrIN
Molecular Weight: 283.89
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 123

Formula: C₅H₃BrIN
MW: 283.89
Melting Pt: 123 °C
MDL Number: MFCD03095201
CAS Number: 73290-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=516c10e839934ead051cdaa1b1ab81eecb6d629b&vid=516c10e839934ead051cdaa1b1ab81eecb6d629b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR