2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3764-25G B3764-5G
TCB3764-25GEA 353.92 CAD
TCB3764-25G TCB3764-5G
2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid ≥98.0% (by GC, titration analysis)
2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid

CAS Number: 4442-54-0
MDL Number: MFCD00463509
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138

Formula: C₉H₈O₄
Melting Pt: 138 °C
MDL Number: MFCD00463509
CAS Number: 4442-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc811abf8161a48d967af16a9752088616969f0a&vid=fc811abf8161a48d967af16a9752088616969f0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR