2-Bromo-4-tert-butylphenol ≥97.0%

Supplier: TCI America

B3763-25G B3763-5G
TCB3763-25GEA 175.22 CAD
TCB3763-25G TCB3763-5G
2-Bromo-4-tert-butylphenol ≥97.0%
2-Bromo-4-tert-butylphenol

CAS Number: 2198-66-5
MDL Number: MFCD02682891
Molecular Formula: C10H13BrO
Molecular Weight: 229.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 113
Melting point (°C): 51

Formula: C₁₀H₁₃BrO
Boiling Pt: 113 °C
Melting Pt: 51 °C
MDL Number: MFCD02682891
CAS Number: 2198-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41eee1c18ea758d03df092adf0193f7e2f47900f&vid=41eee1c18ea758d03df092adf0193f7e2f47900f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR