5-Bromobenzo[b]thiophene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
5-Bromobenzothiophene
B3762-1G
B3762-5G
TCB3762-1GEA
34.76
CAD
TCB3762-1G
TCB3762-5G
5-Bromobenzo[b]thiophene ≥98.0% (by GC)
5-Bromobenzo[b]thiophene
CAS Number: 4923-87-9
MDL Number: MFCD03069318
Molecular Formula: C8H5BrS
Molecular Weight: 213.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 107
Melting point (°C): 49
Formula:
C₈H₅BrS MW: 213.1 g/mol Boiling Pt: 107 °C Melting Pt: 49 °C |
MDL Number:
MFCD03069318 CAS Number: 4923-87-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...