1-Benzhydrylpiperazine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3760-25G
B3760-5G
TCB3760-25GEA
414.91
CAD
TCB3760-25G
TCB3760-5G
1-Benzhydrylpiperazine ≥98.0% (by GC, titration analysis)
1-Benzhydrylpiperazine
CAS Number: 841-77-0
MDL Number: MFCD00038379
Molecular Formula: C17H20N2
Molecular Weight: 252.36
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 93
Flash Point (°C): 115
Other Product Names / Synonyms: 1-(Diphenylmethyl)piperazine
Formula:
C₁₇H₂₀N₂ Boiling Pt: 185 °C Melting Pt: 93 °C Flash Pt: 115 °C |
MDL Number:
MFCD00038379 CAS Number: 841-77-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...