2-Bromo-6-(trifluoromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

B3759-1G B3759-5G
TCB3759-1GEA 30.02 CAD
TCB3759-1G TCB3759-5G
2-Bromo-6-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Bromo-6-(trifluoromethyl)pyridine

CAS Number: 189278-27-1
MDL Number: MFCD00153087
Molecular Formula: C6H3BrF3N
Molecular Weight: 226.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystalline Powder
Boiling point (°C): 96
Melting point (°C): 50

Formula: C₆H₃BrF₃N
MW: 226
Boiling Pt: 96 °C
Melting Pt: 50 °C
MDL Number: MFCD00153087
CAS Number: 189278-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ad6b2706e473eb31af1c2cceb90d7d2ad874531&vid=7ad6b2706e473eb31af1c2cceb90d7d2ad874531&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR