N-(tert-Butoxycarbonyl)-L-prolinamide ≥98.0%

Supplier: TCI America

Synonyms: tert-Butyl (S)-2-Carbamoyl-1-pyrrolidinecarboxylate, (S)-2-Carbamoyl-1-pyrrolidinecarboxylic Acid tert-Butyl Ester, 1-Boc-L-prolinamide, N-Boc-L-prolinamide, Boc-Pro-NH2

B3756-1G B3756-5G
TCB3756-1GEA 42.66 CAD
TCB3756-1G TCB3756-5G
N-(tert-Butoxycarbonyl)-L-prolinamide ≥98.0%
N-(tert-Butoxycarbonyl)-L-prolinamide

CAS Number: 35150-07-3
MDL Number: MFCD00190827
Molecular Formula: C10H18N2O3
Molecular Weight: 214.27
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 107
Specific rotation [a]20/D: -45 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acid Amides
Other Product Names / Synonyms: tert-Butyl (S)-2-Carbamoyl-1-pyrrolidinecarboxylate, (S)-2-Carbamoyl-1-pyrrolidinecarboxylic Acid tert-Butyl Ester, N-Boc-L-prolinamide

Formula: C₁₀H₁₈N₂O₃
MW: 214.27 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00190827
CAS Number: 35150-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4453c3a14f849d2c0718d9030a4113f275d39458&vid=4453c3a14f849d2c0718d9030a4113f275d39458&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR