tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate ≥97.0%

Supplier: TCI America

Synonyms: tert-Butyl 2-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-2-hydroxymethylpiperidine, Tert-Butyl 2-(Hydroxymethyl)Tetrahydro-1(2H)-Pyridinecarboxylate, (±)-1-Boc-2-(hydroxymethyl)piperidine, BOC-2-piperidylmethanol

B3755-25G B3755-5G
TCB3755-25GEA 330.54 CAD
TCB3755-25G TCB3755-5G
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate ≥97.0%
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate

CAS Number: 157634-00-9
MDL Number: MFCD04114966
Molecular Formula: C11H21NO3
Molecular Weight: 215.29
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 81


Other Product Names / Synonyms: 1-Boc-2-piperidinemethanol, 1-(tert-Butoxycarbonyl)-2-(hydroxymethyl)piperidine, 1-Boc-2-(hydroxymethyl)piperidine

Formula: C₁₁H₂₁NO₃
MW: 215.293 g/mol
Melting Pt: 81 °C
MDL Number: MFCD04114966
CAS Number: 157634-00-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd5f350aec972caebd7d8d31a30833903f19e3a6&vid=fd5f350aec972caebd7d8d31a30833903f19e3a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR