N-(tert-Butoxycarbonyl)-L-tert-leucine ≥98.0%
Supplier: TCI America
Synonyms:
N-Boc-L-tert-leucine, Boc-Tle-OH, (S)-2-(Boc-amino)-3,3-dimethylbutyric Acid, (S)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid
CAS Number: 62965-35-9
MDL Number: MFCD00065574
Molecular Formula: C11H21NO4
Molecular Weight: 231.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 119
Specific rotation [a]20/D: -4 deg (C=1, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: (S)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid, (S)-2-(Boc-amino)-3,3-dimethylbutyric Acid, N-Boc-L-tert-leucine, Boc-Tle-OH
Formula:
C₁₁H₂₁NO₄ MW: 231.29 g/mol Melting Pt: 119 °C |
MDL Number:
MFCD00065574 CAS Number: 62965-35-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...