Bis(2-bromoethyl)ether ≥98.0%
Supplier: TCI America
Synonyms:
1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether
B3752-25G
B3752-5G
TCB3752-25GEA
143.78
CAD
TCB3752-25G
TCB3752-5G
Bis(2-bromoethyl)ether ≥98.0%
Bis(2-bromoethyl) ether
CAS Number: 5414-19-7
MDL Number: MFCD00039196
Molecular Formula: C4H8Br2O
Molecular Weight: 231.92
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 45
Flash Point (°C): 85
Specific Gravity (20/20): 1.86
Other Product Names / Synonyms: 2,2'-Dibromodiethyl Ether, 2-Bromoethyl Ether
Formula:
C₄H₈Br₂O Boiling Pt: 45 °C Density: 1.86 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00039196 CAS Number: 5414-19-7 UN: 2810 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...