5-Bromo-2-chloro-3-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-2-chloro-3-picoline

B3744-25G B3744-5G
TCB3744-25GEA 279.34 CAD
TCB3744-25G TCB3744-5G
5-Bromo-2-chloro-3-methylpyridine ≥98.0%
5-Bromo-2-chloro-3-methylpyridine

CAS Number: 29241-60-9
MDL Number: MFCD03095093
Molecular Formula: C6H5BrClN
Molecular Weight: 206.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 78
Melting point (°C): 44


Other Product Names / Synonyms: 5-Bromo-2-chloro-3-picoline

Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 78 °C
Melting Pt: 44 °C
MDL Number: MFCD03095093
CAS Number: 29241-60-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f84e65dbe3ebf0867be208e620c64602ae7c6f7&vid=6f84e65dbe3ebf0867be208e620c64602ae7c6f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR