4-Bromo-3-fluoroanisole ≥98.0% (by GC)

Supplier: TCI America

B3743-25G B3743-5G
TCB3743-25GEA 371.3 CAD
TCB3743-25G TCB3743-5G
4-Bromo-3-fluoroanisole ≥98.0% (by GC)
4-Bromo-3-fluoroanisole

CAS Number: 458-50-4
MDL Number: MFCD01310983
Molecular Formula: C7H6BrFO
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Specific Gravity (20/20): 1.60

Formula: C₇H₆BrFO
Boiling Pt: 216 °C
Density: 1.60 (20 °C)
MDL Number: MFCD01310983
CAS Number: 458-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63e4e4486890d344ae96f56e0f4ef47132bc077f&vid=63e4e4486890d344ae96f56e0f4ef47132bc077f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR