5-Bromooxindole ≥97.0%

Supplier: TCI America

Synonyms: 5-Bromo-1,3-dihydro-2H-indol-2-one, 5-Bromoindolin-2-one, 5-Bromo-2-oxindole

B3741-1G B3741-5G
TCB3741-1GEA 66.04 CAD
TCB3741-1G TCB3741-5G
5-Bromooxindole ≥97.0%
5-Bromooxindole

CAS Number: 20870-78-4
MDL Number: MFCD00456998
Molecular Formula: C8H6BrNO
Molecular Weight: 212.05
Purity/Analysis Method: >97.0% (GC,N)
Form: Crystal
Melting point (°C): 222


Other Product Names / Synonyms: 5-Bromo-2-indolinone

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00456998
CAS Number: 20870-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2319a3a207d8a3bce2f9e4b227b5fafbb9b8e68&vid=b2319a3a207d8a3bce2f9e4b227b5fafbb9b8e68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR