4-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
B3739-1G
B3739-5G
TCB3739-1GEA
80.58
CAD
TCB3739-1G
TCB3739-5G
4-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(tert-Butyldimethylsilyloxy)phenylboronic acid
CAS Number: 159191-56-7
MDL Number: MFCD03093888
Molecular Formula: C12H21BO3Si
Molecular Weight: 252.19
Form: Crystal
Color: White
Melting point (°C): 207
Other Product Names / Synonyms: 4-(tert-Butyldimethylsilyloxy)benzeneboronic Acid
Formula:
C₁₂H₂₁BO₃Si Melting Pt: 207 °C |
MDL Number:
MFCD03093888 CAS Number: 159191-56-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...