2-Bromo-5-n-octylthiophene ≥97.0% (by GC)
Supplier: TCI America
B3738-25G
B3738-5G
TCB3738-25GEA
1095.89
CAD
TCB3738-25G
TCB3738-5G
2-Bromo-5-n-octylthiophene ≥97.0% (by GC)
2-Bromo-5-n-octylthiophene
CAS Number: 172514-63-5
Molecular Formula: C12H19BrS
Molecular Weight: 275.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Melting point (°C): 5
Storage Temperature: 0-10°C
Formula:
C₁₂H₁₉BrS Boiling Pt: 112 °C Melting Pt: 5 °C |
CAS Number:
172514-63-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...