3-Bromo-2-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3737-1G B3737-5G
TCB3737-1GEA 63.2 CAD
TCB3737-1G TCB3737-5G
3-Bromo-2-methoxypyridine ≥98.0% (by GC, titration analysis)
3-Bromo-2-methoxypyridine

CAS Number: 13472-59-8
MDL Number: MFCD03095349
Molecular Formula: C6H6BrNO
Molecular Weight: 188.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 93
Specific Gravity (20/20): 1.58

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Density: 1.58 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD03095349
CAS Number: 13472-59-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=306d7bc59054088b269c8e9d275b990f96dd8778&vid=306d7bc59054088b269c8e9d275b990f96dd8778&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR