tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥98.0% (by GC)
Supplier: TCI America
B3735-25G
B3735-5G
TCB3735-25GEA
426.6
CAD
TCB3735-25G
TCB3735-5G
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥98.0% (by GC)
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate
CAS Number: 73286-70-1
MDL Number: MFCD01863512
Molecular Formula: C9H15NO2
Molecular Weight: 169.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 75
Melting point (°C): 41
Flash Point (°C): 81
Other Product Names / Synonyms: N-Boc-3-pyrroline
Formula:
C₉H₁₅NO₂ Boiling Pt: 75 °C Melting Pt: 41 °C Flash Pt: 81 °C |
MDL Number:
MFCD01863512 CAS Number: 73286-70-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...