2-Bromo-3,5-dichloropyridine ≥98.0% (by GC)
Supplier: TCI America
B3734-25G
B3734-5G
TCB3734-25GEA
206.98
CAD
TCB3734-25G
TCB3734-5G
2-Bromo-3,5-dichloropyridine ≥98.0% (by GC)
2-Bromo-3,5-dichloropyridine
CAS Number: 14482-51-0
MDL Number: MFCD00233990
Molecular Formula: C5H2BrCl2N
Molecular Weight: 226.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 43
Formula:
C₅H₂BrCl₂N Melting Pt: 43 °C |
MDL Number:
MFCD00233990 CAS Number: 14482-51-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...