6-Bromo-4,4'-dimethyl-2,2'-bipyridyl ≥97.0%
Supplier: TCI America
Synonyms:
6-Bromo-4,4'-dimethyl-2,2'-bipyridine
B3732-1G
B3732-5G
TCB3732-1GEA
388.68
CAD
TCB3732-1G
TCB3732-5G
6-Bromo-4,4'-dimethyl-2,2'-bipyridyl ≥97.0%
6-Bromo-4,4'-dimethyl-2,2'-bipyridyl
CAS Number: 850413-36-4
MDL Number: MFCD11042495
Molecular Formula: C12H11BrN2
Molecular Weight: 263.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 107
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 6-Bromo-4,4'-dimethyl-2,2'-bipyridine
Formula:
C₁₂H₁₁BrN₂ MW: 263.14 g/mol Melting Pt: 107 °C |
MDL Number:
MFCD11042495 CAS Number: 850413-36-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...