5-Bromoresorcinol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3731-1G B3731-5G
TCB3731-1GEA 218.04 CAD
TCB3731-1G TCB3731-5G
5-Bromoresorcinol ≥98.0% (by GC, titration analysis)
5-Bromoresorcinol

CAS Number: 106120-04-1
MDL Number: MFCD16293393
Molecular Formula: C6H5BrO2
Molecular Weight: 189.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 93


Other Product Names / Synonyms: 1-Bromo-3,5-dihydroxybenzene

Formula: C₆H₅BrO₂
Boiling Pt: 140 °C
Melting Pt: 93 °C
MDL Number: MFCD16293393
CAS Number: 106120-04-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6696a007d36cc5a9530205510030f75806c66c42&vid=6696a007d36cc5a9530205510030f75806c66c42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR