4-Bromo-2-methyl-6-nitroaniline ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene

B3730-1G B3730-5G
TCB3730-1GEA 39.5 CAD
TCB3730-1G TCB3730-5G
4-Bromo-2-methyl-6-nitroaniline ≥98.0%
4-Bromo-2-methyl-6-nitroaniline

CAS Number: 77811-44-0
MDL Number: MFCD00052919
Molecular Formula: C7H7BrN2O2
Molecular Weight: 231.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 146


Other Product Names / Synonyms: 4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene

Formula: C₇H₇BrN₂O₂
Melting Pt: 146 °C
MDL Number: MFCD00052919
CAS Number: 77811-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c83c13100080301f1338dc80fe5c56c9e13e3a15&vid=c83c13100080301f1338dc80fe5c56c9e13e3a15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR