3-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-(tert-Butyl dimethylsiloxy)phenyl boronic acid, (3-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid, 3-(tert-Butyldimethylsiloxy)benzeneboronic acid

B3726-1G B3726-5G
TCB3726-1GEA 91.64 CAD
TCB3726-1G TCB3726-5G
3-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)
3-(tert-Butyldimethylsilyloxy)phenylboronic acid

CAS Number: 261621-12-9
MDL Number: MFCD03701505
Molecular Formula: C12H21BO3Si
Molecular Weight: 252.19
Form: Crystal
Color: White
Melting point (°C): 91


Other Product Names / Synonyms: 3-(tert-Butyldimethylsilyloxy)benzeneboronic Acid

Formula: C₁₂H₂₁BO₃Si
MW: 252.19 g/mol
Melting Pt: 91 °C
MDL Number: MFCD03701505
CAS Number: 261621-12-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7297c659eb7ea9e20a9bfff871d98eb8d6c34cf7&vid=7297c659eb7ea9e20a9bfff871d98eb8d6c34cf7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR