3-Bromo-5-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3724-1G B3724-5G
TCB3724-1GEA 67.94 CAD
TCB3724-1G TCB3724-5G
3-Bromo-5-methylpyridine ≥98.0% (by GC, titration analysis)
3-Bromo-5-methylpyridine

CAS Number: 3430-16-8
MDL Number: MFCD01646141
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 91
Freezing point (°C): 19
Specific Gravity (20/20): 1.52

Formula: C₆H₆BrN
Boiling Pt: 110 °C
Density: 1.52 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD01646141
CAS Number: 3430-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=940478d5b7f425863a7c710cc3983c399b246ae1&vid=940478d5b7f425863a7c710cc3983c399b246ae1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR