2-Bromo-3-fluorobenzoic acid ≥96.0% (by GC, titration analysis)
Supplier: TCI America
B3722-1G
B3722-5G
TCB3722-1GEA
115.66
CAD
TCB3722-1G
TCB3722-5G
2-Bromo-3-fluorobenzoic acid ≥96.0% (by GC, titration analysis)
2-Bromo-3-fluorobenzoic acid
CAS Number: 132715-69-6
MDL Number: MFCD01569398
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 160
Formula:
C₇H₄BrFO₂ Melting Pt: 160 °C |
MDL Number:
MFCD01569398 CAS Number: 132715-69-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...