3-Bromo-2-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America

B3721-1G B3721-5G
TCB3721-1GEA 28.44 CAD
TCB3721-1G TCB3721-5G
3-Bromo-2-fluoropyridine ≥98.0% (by GC)
3-Bromo-2-fluoropyridine

CAS Number: 36178-05-9
MDL Number: MFCD04112496
Molecular Formula: C5H3BrFN
Molecular Weight: 175.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 76
Specific Gravity (20/20): 1.73

Formula: C₅H₃BrFN
Boiling Pt: 76 °C
Density: 1.73 (20 °C)
MDL Number: MFCD04112496
CAS Number: 36178-05-9
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a6753e6f47650a1b267a581fb47cc31fd0193eb&vid=4a6753e6f47650a1b267a581fb47cc31fd0193eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR