tert-Butyl ethyl malonate ≥95.0% (by GC)
Supplier: TCI America
B3720-25G
B3720-5G
TCB3720-25GEA
164.32
CAD
TCB3720-25G
TCB3720-5G
tert-Butyl ethyl malonate ≥95.0% (by GC)
tert-Butyl ethyl malonate
CAS Number: 32864-38-3
MDL Number: MFCD00009193
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 87
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: Malonic Acid tert-Butyl Ethyl Ester
Formula:
C₉H₁₆O₄ Boiling Pt: 84 °C Density: 1.00 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00009193 CAS Number: 32864-38-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...