2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol ≥98.0% (by HPLC)

Supplier: TCI America

B3717-250G B3717-25G
TCB3717-250GEA 197.5 CAD
TCB3717-250G TCB3717-25G
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol ≥98.0% (by HPLC)
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol

CAS Number: 70321-86-7
MDL Number: MFCD00134705
Molecular Formula: C30H29N3O
Molecular Weight: 447.58
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 140
Flash Point (°C): 195

Formula: C₃₀H₂₉N₃O
Melting Pt: 140 °C
Flash Pt: 195 °C
MDL Number: MFCD00134705
CAS Number: 70321-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45ab65fc28f88649f626b12a0fdc4171ce4285ad&vid=45ab65fc28f88649f626b12a0fdc4171ce4285ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR