2-Bromo-4-formylpyridine ≥95.0%

Supplier: TCI America

Synonyms: 2-Bromo-4-pyridinecarboxaldehyde, 2-Bromopyridine-4-carboxaldehyde

B3715-1G B3715-5G
TCB3715-1GEA 39.5 CAD
TCB3715-1G TCB3715-5G
2-Bromo-4-formylpyridine ≥95.0%
2-Bromo-4-formylpyridine

CAS Number: 118289-17-1
MDL Number: MFCD04039311
Molecular Formula: C6H4BrNO
Molecular Weight: 186.01
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 55
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Bromo-4-formylpyridine, 2-Bromoisonicotinaldehyde

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 55 °C
MDL Number: MFCD04039311
CAS Number: 118289-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ef44c4e23c3bfff3bba79947b97280e9307d9b4&vid=9ef44c4e23c3bfff3bba79947b97280e9307d9b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR