2-Bromo-3-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America

B3712-1G B3712-5G
TCB3712-1GEA 135.88 CAD
TCB3712-1G TCB3712-5G
2-Bromo-3-nitrotoluene ≥98.0% (by GC)
2-Bromo-3-nitrotoluene

CAS Number: 41085-43-2
MDL Number: MFCD00134555
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 40
Flash Point (°C): 113

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
Boiling Pt: 136 °C
Melting Pt: 40 °C
Flash Pt: 113 °C
MDL Number: MFCD00134555
CAS Number: 41085-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e44f8e91a94bd26e1084614ad33be5f9967893&vid=a0e44f8e91a94bd26e1084614ad33be5f9967893&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR