6-Bromo-1-indanone ≥98.0% (by GC)

Supplier: TCI America

B3711-1G B3711-5G
TCB3711-1GEA 55.3 CAD
TCB3711-1G TCB3711-5G
6-Bromo-1-indanone ≥98.0% (by GC)
6-Bromo-1-indanone

CAS Number: 14548-39-1
MDL Number: MFCD02179286
Molecular Formula: C9H7BrO
Molecular Weight: 211.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 112
Lambda max.: 302 nm (EtOH)

Formula: C₉H₇BrO
MW: 211.06 g/mol
Melting Pt: 112 °C
MDL Number: MFCD02179286
CAS Number: 14548-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b3fcdf792ae7fe3145e9d3e0421906b4adc315b&vid=3b3fcdf792ae7fe3145e9d3e0421906b4adc315b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR