3-Bromocatechol ≥98.0% (by GC)

Supplier: TCI America

B3710-1G B3710-5G
TCB3710-1GEA 91.64 CAD
TCB3710-1G TCB3710-5G
3-Bromocatechol ≥98.0% (by GC)
3-Bromocatechol

CAS Number: 14381-51-2
MDL Number: MFCD00869768
Molecular Formula: C6H5BrO2
Molecular Weight: 189.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 43


Other Product Names / Synonyms: 1-Bromo-2,3-dihydroxybenzene

Formula: C₆H₅BrO₂
Boiling Pt: 120 °C
Melting Pt: 43 °C
MDL Number: MFCD00869768
CAS Number: 14381-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45bf52638ce08cbd9c2c7b16fb2025bc7bd5e578&vid=45bf52638ce08cbd9c2c7b16fb2025bc7bd5e578&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR