4-Bromo-3-chloroaniline ≥98.0%

Supplier: TCI America

B3709-25G B3709-5G
TCB3709-25GEA 571.96 CAD
TCB3709-25G TCB3709-5G
4-Bromo-3-chloroaniline ≥98.0%
4-Bromo-3-chloroaniline

CAS Number: 21402-26-6
MDL Number: MFCD00041311
Molecular Formula: C6H5BrClN
Molecular Weight: 206.47
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 66

Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 264
Melting Pt: 66 °C
MDL Number: MFCD00041311
CAS Number: 21402-26-6
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32bdd5a9a25436c743fb2db578e5e90bcb7b2e4c&vid=8346432f9e1d7f0fb76cb06b3ff1f5c9b1ee5498&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR