1-Boc-4-piperidineacetic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: [1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid, 2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid, 1-(tert-Butoxycarbonyl)-4-piperidylacetic acid

B3703-1G B3703-5G
TCB3703-1GEA 56.88 CAD
TCB3703-1G TCB3703-5G
1-Boc-4-piperidineacetic acid ≥97.0% (by GC, titration analysis)
1-Boc-4-piperidineacetic acid

CAS Number: 157688-46-5
MDL Number: MFCD00800239
Molecular Formula: C12H21NO4
Molecular Weight: 243.30
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 99


Other Product Names / Synonyms: 1-Boc-4-piperidylacetic Acid

Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00800239
CAS Number: 157688-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=145f35c38efb23bbf2d6c2ac6d6cc78ddcd20f04&vid=145f35c38efb23bbf2d6c2ac6d6cc78ddcd20f04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR