(4R,6R)-tert-Butyl-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetate ≥98.0%
Supplier: TCI America
Synonyms:
tert-Butyl 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate, 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetic acid tert-Butyl ester
CAS Number: 125995-13-3
MDL Number: MFCD07369252
Molecular Formula: C14H27NO4
Molecular Weight: 273.37
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: 15 deg (C=1, CHCl3)
Lambda max.: 204 nm (EtOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: 2-[(4R,6R)-6-(2-Aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetic Acid tert-Butyl Ester
Formula:
C14H27NO4 MW: 273.3700 g/mol Density: 1.02 (20 °C) |
MDL Number:
MFCD07369252 CAS Number: 125995-13-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...