(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-Homopro-OH, 1-(tert-Butoxycarbonyl)-L-pipecolic Acid, Boc-HomoPro-OH, 1-Boc-L-pipecolic Acid, (S)-1-Boc-2-piperidinecarboxylic Acid, Boc-Pip-OH, (S)-N-BOC-Piperidine-2-carboxylic acid

B3700-1G B3700-5G
TCB3700-1GEA 53.72 CAD
TCB3700-1G TCB3700-5G
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid ≥98.0%
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

CAS Number: 26250-84-0
MDL Number: MFCD00151904
Molecular Formula: C11H19NO4
Molecular Weight: 229.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 124
Specific rotation [a]20/D: -46 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-(tert-Butoxycarbonyl)-L-pipecolic Acid, 1-Boc-L-pipecolic Acid, Boc-Pip-OH, (S)-1-Boc-2-piperidinecarboxylic Acid

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 124 °C
MDL Number: MFCD00151904
CAS Number: 26250-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59030fad76bfcc9e8b934cc332b5c97f5cd369e&vid=f59030fad76bfcc9e8b934cc332b5c97f5cd369e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR