2-Bromo-3-dodecylthiophene ≥97.0% (by GC)
Supplier: TCI America
B3691-5G
B3691-1G
TCB3691-5GEA
419.81
CAD
TCB3691-5G
TCB3691-1G
2-Bromo-3-dodecylthiophene ≥97.0% (by GC)
2-Bromo-3-dodecylthiophene
CAS Number: 139100-06-4
MDL Number: MFCD10000891
Molecular Formula: C16H27BrS
Molecular Weight: 331.36
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 110
Specific Gravity (20/20): 1.13
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-Bromo-3-laurylthiophene
Formula:
C₁₆H₂₇BrS Boiling Pt: 165 °C Density: 1.13 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD10000891 CAS Number: 139100-06-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...