4-Bromo-3-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3689-25G B3689-5G
TCB3689-25GEA 420.28 CAD
TCB3689-25G TCB3689-5G
4-Bromo-3-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-3-nitrobenzoic acid

CAS Number: 6319-40-0
MDL Number: MFCD00272137
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 204

Formula: C₇H₄BrNO₄
Melting Pt: 204 °C
MDL Number: MFCD00272137
CAS Number: 6319-40-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26b80f8740cf7e9d19f437ea41719fcdddde5c29&vid=26b80f8740cf7e9d19f437ea41719fcdddde5c29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR