4-Bromo-3-fluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3688-25G B3688-5G
TCB3688-25GEA 619.36 CAD
TCB3688-25G TCB3688-5G
4-Bromo-3-fluorophenol ≥98.0% (by GC, titration analysis)
4-Bromo-3-fluorophenol

CAS Number: 121219-03-2
MDL Number: MFCD00051907
Molecular Formula: C6H4BrFO
Molecular Weight: 191.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 73

Formula: C₆H₄BrFO
Boiling Pt: 96 °C
Melting Pt: 73 °C
MDL Number: MFCD00051907
CAS Number: 121219-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca8f6331145ee9a5f7ca619e765cd29c97451f75&vid=ca8f6331145ee9a5f7ca619e765cd29c97451f75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR