5-Bromo-o-toluidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B3687-25G B3687-5G
TCB3687-25GEA 184.86 CAD
TCB3687-25G TCB3687-5G
5-Bromo-o-toluidine ≥97.0% (by GC, titration analysis)
5-Bromo-o-toluidine

CAS Number: 39478-78-9
MDL Number: MFCD00800678
Molecular Formula: C7H8BrN
Molecular Weight: 186.05
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 139
Flash Point (°C): 110
Freezing point (°C): 30


Other Product Names / Synonyms: 2-Amino-4-bromotoluene, 5-Bromo-o-toluidine

Formula: C₇H₈BrN
Boiling Pt: 139 °C
Flash Pt: 110 °C
MDL Number: MFCD00800678
CAS Number: 39478-78-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14e986fd6ef2267efb51b5a6e336a82a0879100f&vid=14e986fd6ef2267efb51b5a6e336a82a0879100f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR