2-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America

B3686-25G B3686-5G
TCB3686-25GEA 151.68 CAD
TCB3686-25G TCB3686-5G
2-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98.0% (by GC)
2-Bromo-α,α,α-trifluoro-5-nitrotoluene

CAS Number: 367-67-9
MDL Number: MFCD00014707
Molecular Formula: C7H3BrF3NO2
Molecular Weight: 270.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 88
Melting point (°C): 44
Flash Point (°C): 113


Other Product Names / Synonyms: 1-Bromo-4-nitro-2-(trifluoromethyl)benzene

Formula: C₇H₃BrF₃NO₂
Boiling Pt: 88 °C
Melting Pt: 44 °C
Flash Pt: 113 °C
MDL Number: MFCD00014707
CAS Number: 367-67-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2436770ece19f39b0ae3b3b35f6e722c31c29692&vid=2436770ece19f39b0ae3b3b35f6e722c31c29692&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR